Anti-FAM212A Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB94351-50
BIRBORB94351-50EA 0 EUR
BIRBORB94351-50
Anti-FAM212A Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 5 ?g/ml; Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: FAM212A (family with sequence similarity 212, member A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR