Anti-GM2A Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB94808-50
BIRBORB94808-50EA 0 EUR
BIRBORB94808-50
Anti-GM2A Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 10-12.5 ?g/ml

Type: Primary
Antigen: GM2A (GM2 ganglioside activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR