Anti-RGS19 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB95735-50
BIRBORB95735-50EA 0 EUR
BIRBORB95735-50
Anti-RGS19 Rabbit Polyclonal Antibody
Antibodies
Immunochemistry: 10 ?g/ml; Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: RGS19 (regulator of G-protein signaling 19)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR