Anti-STOM Goat Polyclonal Antibody

Supplier: Biorbyt
ORB97643-100
BIRBORB97643-100EA 0 EUR
BIRBORB97643-100
Anti-STOM Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: STOM (stomatin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR