Cleanroom gloves, nitrile, BioClean Nerva™
Supplier: NITRITEX
BNAL-L BNAL-M BNAL-S BNAL-XL BNAL-XXL
BNAL-LEA 940 EUR
BNAL-L BNAL-M BNAL-S BNAL-XL 112-3852
Cleanroom gloves, nitrile, BioClean Nerva™
Gloves
These ambidextrous nitrile gloves with beaded cuffs provide good resistance against chemicals. They also have excellent ESD properties.
  • Non sterile, powder-free, textured fingertips
  • Contain no natural latex proteins, hypoallergenic
  • Extended length provides protection of the forearm
  • Extended leaching in pure deionised water ensures extremely low levels of chlorides and extractable metals
  • Textured fingers provide enhanced grip
Suitable for use in Class 10, ISO 4 or M2.5 cleanrooms.
Certifications: Manufactured in a facility holding ISO 9001:2015 and (More Product Information