Anti-KDR Mouse Monoclonal Antibody (CF405S) [clone: KDR657]
Supplier: Biotium
BNC040657-500 BNC040657-100
BNC040657-500EA 560 EUR
BNC040657-500 BNC040657-100
Anti-KDR Mouse Monoclonal Antibody (CF405S) [clone: KDR657]
Antibodies
CD309, also known as VEGF-R2, KDR3, and Flk-1 (mouse), is a type I transmembrane glycoprotein. It is a member of the CSF-1/PDGF receptor family of type III tyrosine kinase receptors. Human VEGF-R2 is mainly expressed by endothelial cells, embryonic tissues, and megakaryocytes. It plays an important role in the regulation of angiogenesis, vasculogenesis, and vascular permeability. The ligands of VEGF-R2 include VEGF-A, VEGF-C, VEGF-D, and VEGF splice isoforms. Ligation of VEGF-R2 with its ligands results in the receptor dimerization and auto-phosphorylation, stimulating endothelial cellMore Product Information