Anti-NEFH Mouse Monoclonal Antibody (CF640R) [clone: RT-97]
Supplier: Biotium
BNC400454-500 BNC400454-100
BNC400454-500EA 560 EUR
BNC400454-500 BNC400454-100
Anti-NEFH Mouse Monoclonal Antibody (CF640R) [clone: RT-97]
Antibodies
This MAb reacts with a 200 kDa protein, identified as heavy sub-unit of neurofilaments (NF-H). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68 kDa (NF-L), 160 kDa (NF-M) and 200 kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments areMore Product Information