Anti-MUC1 Mouse Monoclonal Antibody (CF647) [clone: MUC1/845]
Supplier: Biotium
BNC470845-500 BNC470845-100
BNC470845-500EA 560 EUR
BNC470845-500 BNC470845-100
Anti-MUC1 Mouse Monoclonal Antibody (CF647) [clone: MUC1/845]
Antibodies
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. The α subunit has cell adhesive properties. It can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The β subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinoma tissues. Antibody to EMA is useful as a pan-More Product Information