Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF488A) [clone: AE-1]
Supplier: Biotium
BNC880905-500 BNC880905-100
BNC880905-500EA 560 EUR
BNC880905-500 BNC880905-100
Anti-Mitochondrial Marker Mouse Monoclonal Antibody (CF488A) [clone: AE-1]
Antibodies
This MAb recognizes a 60 kDa antigen associated with the mitochondria in human cells. It is a part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. It recognizes an antigen associated with the mitochondria in human cells only. It can be used to stain the mitochondria in cell or tissue preparations and can be used as a mitochondrial marker in subcellular fractions. It produces a spaghetti-like pattern in normal and malignantMore Product Information