Anti-CD8A Mouse Monoclonal Antibody (CF488A) [clone: C8/1035]
Supplier: Biotium
BNC881035-500 BNC881035-100
BNC881035-500EA 560 EUR
BNC881035-500 BNC881035-100
Anti-CD8A Mouse Monoclonal Antibody (CF488A) [clone: C8/1035]
Antibodies
CD8 is a cell surface receptor expressed either as a heterodimer with the CD8 β chain (CD8 α/β) or as a homodimer (CD8 α/α). A majority of thymocytes and a subpopulation of mature T cells and NK cells express CD8a. CD8 binds to MHC class 1 and through its association with protein tyrosine kinase p56lck plays a role in T cell development and activation of mature T cells. For mature T-cells, CD4 and CD8 are mutually exclusive, so anti-CD8, generally used in conjunction with anti-CD4. It is a useful marker for distinguishing helper/inducer T-lymphocytes, and most peripheral T-cell lymphomas areMore Product Information