Anti-CD14 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: MDA/929]

Supplier: Biotium
BNUM0929-50
BNUM0929-50EA 417 EUR
BNUM0929-50
Anti-CD14 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: MDA/929]
Antibodies
Recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MDA/929
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol CD14
Environmentally Preferable
Antigen synonyms Monocyte Differentiation Antigen 14|LPSR|Mo2|Myeloid cell-specific leucine-rich glycoprotein|LPS-R|Lipopolysaccharide receptor
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone Clone MDA/929
Reactivity Human
Host Mouse
Gene ID 163867
Isotype IgG1 kappa
ImmunoFluorescence Yes
Purification BSA-free
Concentration 1 mg/mL
Storage buffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR