Double bosshead, Jumbo
Supplier: BOCHEM
5650
BOCH5650EA 96.6 EUR
BOCH5650
Double bosshead, Jumbo
Clamps Bossheads
Double bossheads made of chrome plated aluminium.
  • For rods up to 20/30 mm
  • Angle: 90°
  • Thread: 10