Pots, laboratory, without lid
Supplier: BOCHEM
8363 8367 8373 8377 8362 8366 8364 8365 8370 8360 8372 8374 8375 8376 8371 8361
BOCH8363EA 227 EUR
BOCH8363 BOCH8367 BOCH8373 BOCH8377 BOCH8362 BOCH8366 BOCH8364 BOCH8365 BOCH8370 BOCH8360 BOCH8372 BOCH8374 BOCH8375 BOCH8376 BOCH8371 BOCH8361
Pots, laboratory, without lid
Trays/Dishes/Pots Pots
Stainless steel 18/10.
  • Rounded corners for fast, easy cleaning
  • Good chemical resistance
  • Very good thermal resistance
Multipurpose pots for a wide range of uses. This series is suitable for induction fields.
Ordering information: Lids to be ordered separately.