Gas washing bottles, micro scrubber, BOLA
Supplier: Bohlender
A119-24
BOHLA119-24EA 541 EUR
BOHLA119-24
Gas washing bottles, micro scrubber, BOLA
Bottles Gas Washing Bottles
PFA, translucent.
  • Used as cold trap
  • Temperature resistance from –200 to +250 °C
  • Universal chemical resistance
  • Unbreakable and transparent
For use with volumes up to 50 ml. The gas inlet tube (not included, please choose a hard-walled tubing) is inserted through the fitting on the top and can be fixed safely. The side fitting is used as gas outlet.
Certifications: FDA conform.
Order Now
SPECIFICATIONS
Environmentally Preferable