Stirrer bearings, PTFE, BOLA
Supplier: Bohlender
B288-02
BOHLB288-02EA 98.3 EUR
BOHLB288-02
Stirrer bearings, PTFE, BOLA
Stirrers Stirring Tools
PTFE stirrer bearings with adjustable special gasket.
  • Chemical resistance
  • Temperature resistance: −20 to 250 °C
Integrated special nut is present for easy locking and unlocking of the ground joint.