Connectors, for GL threaded connections
Supplier: Bohlender
D584-18 D584-14 D583-25 D583-18 D583-14
201-0271EA 5.5 EUR
201-0271 BOHLD584-14 BOHLD583-25 BOHLD583-18 BOHLD583-14
Connectors, for GL threaded connections
Fittings and Connectors
Connector made from PP with elastic sealing lip and FPM O-ring
  • Good chemical resistance
  • Temperature resistant from −20 to +110°C
  • Suitable for vacuum applications
For use with GL screw caps.