Double tube end fittings, BOLA
Supplier: Bohlender
F703-04 F703-02
BOHLF703-04EA 171 EUR
BOHLF703-04 BOHLF703-02
Double tube end fittings, BOLA
Fixings Screws
PTFE, white.
  • Temperature resistance: from -200 to +250 °C
  • Universal chemical resistance
  • FDA conform
With two male threads UNF ¼" 28 G.