Screw caps for bottles with GL threads, BOLA
Supplier: Bohlender
H986-04 H986-02 H986-03 H986-05 H986-01
BOHLH986-04EA 48.8 EUR
BOHLH986-04 BOHLH986-02 BOHLH986-03 BOHLH986-05 BOHLH986-01
Screw caps for bottles with GL threads, BOLA
Closures Bottle Closures
Screw cap with handy knurl, made of PTFE.
  • Very good chemical resistance
  • Temperature resistant from −200 to +250 °C
Suitable for bottles and vessels with GL threads.
Order Now
SPECIFICATIONS
Environmentally Preferable