Filter disc, BOLA
Supplier: Bohlender
N1565-12
BOHLN1565-12EA 140 EUR
BOHLN1565-12
Filter disc, BOLA
Filters Filter Discs
Filter disc made of porous PTFE.
Temperature resistance: –200 to 250 °C
Certifications: Conforms to FDA.