b.safe Desktop Distributors
Supplier: Bohlender
R1122-04 R1123-02 R1123-04 R1122-02
BOHLR1122-04EA 276 EUR
BOHLR1122-04 BOHLR1123-02 BOHLR1123-04 BOHLR1122-02
b.safe Desktop Distributors
Fittings and Connectors
Central waste collection from several HPLC systems. Capillaries and hoses are connected without kinks using the b.safe Fittings and b.safe Hose connectors included in the scope of delivery. The liquid solvent waste flows off directly and residue-free via the vertical hole inside the Desktop Distributor.
  • Central hose connection
  • Simple attachment to the work table
  • Residue-free drainage of solvent waste
Base plate made of PP, for collecting solvent waste from up to eight capillaries and four hoses and forwarding it via a central hose connection. With PFA-More Product Information