Multicollector, b.safe
Supplier: Bohlender
R1140-03 R1140-04
BOHLR1140-03EA 172 EUR
BOHLR1140-03 BOHLR1140-04
Multicollector, b.safe
Fittings and Connectors
Combination of capillaries and tubes of several HPLC systems in one desktop mount.
  • With a b.safe Multicollector you simply multiply the number of connectors of a b.safe Desktop Mount
  • This extension can be done without any modification of the existing installation
  • Chemically inert
  • Static dissipative to prevent static charging
Made of static dissipative PE-EX, screw cap made of PPS with GL 45 thread, rotary insert of static dissipative PE-EX and male threads GL 45. To connect up to 4 b.safe System Caps GL 45 to one desktop mount GL 45 (not included in scope of delivery).