System caps GL 45, b.safe
Supplier: Bohlender
R1145-02 R1145-03 R1145-04 R1145-05 R1145-07 R1145-14 R1145-16
BOHLR1145-02EA 79.2 EUR
BOHLR1145-02 BOHLR1145-03 BOHLR1145-04 BOHLR1145-05 BOHLR1145-07 BOHLR1145-14 BOHLR1145-16
System caps GL 45, b.safe
Closures Safety Caps and Safety Waste Caps
Screw cap made of PPS for free movable GL 45 thread, insert made of PTFE. With PFA fittings for capillaries with Ext.Ø 1,6/2,2/3,2 mm (thread UNF ¼"), hose connector for tubings Int.Ø 6,5 to 8 mm (thread NPT ¹/₈"). Adapted to the installation on b.safe Desktop Mounts GL 45 or b.safe Multiports GL 45.
  • Perfect protection from hazardous vapours
  • Closed system: Reduction of expensive solvent loss
  • Guarantee of pure eluents
  • Huge choice of cap sizes for compatibility with any eluent flask
  • No twisting of capillaries
  • Screw caps can individually be labelled by hand
More Product Information