Blind cap for waste pipes, b.safe
Supplier: Bohlender
R1160-05
BOHLR1160-05EA 21.3 EUR
BOHLR1160-05
Blind cap for waste pipes, b.safe
Closures Safety Caps and Safety Waste Caps
Cap made of PE-EX with pressed-in conical sealing plate made of static-dissipative PTFE-EX.
Suitable sealing cap for clamp connections for tube Ext.Ø 32 mm.