Safety spectacles, Prism
Supplier: Bollé Safety
PRIPSI
BOLEPRIPSIEA 7.26 EUR
BOLEPRIPSI
Safety spectacles, Prism
Safety Glasses
The model Prism with dual lens offers a high level of protection and its frame is entirely composed of ultra resistant PC.
  • Panoramic vision
  • Sporty design and provides upper protection
  • Comfortable with straight temples
Weight: 23 g
Certifications: EN 166; EN 170
Delivery information: Supplied with adjustable cord.