Anti-CALCA Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-0063R-CY5
BOSSBS-0063R-CY5EA 893 EUR
BOSSBS-0063R-CY5
Anti-CALCA Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Calcitonin causes a rapid but short-lived drop in the level of calcium and phosphate in blood by promoting the incorporation of those ions in the bones. Katacalcin is a potent plasma calcium-lowering peptide.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CALCA
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR