Anti-PADI4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-0318R-HRP
BOSSBS-0318R-HRPEA 900 EUR
BOSSBS-0318R-HRP
Anti-PADI4 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
PADI4 (peptidyl arginine deiminase, type IV) catalyzes the deimination of arginine residues of proteins. Down-regulates histone H3 and H4 arginine methylation, both by preventing arginine methylation by CARM1 and HRMT1L2/PRMT1 and by converting methylarginine to citrulline. subcellular location at cytoplasmic granules. Belongs to the protein arginine deiminase family. This gene may play a role in granulocyte and macrophage development leading to inflammation and immune response.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: PADI4
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR