Anti-Streptavidin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-0437R-A750
BOSSBS-0437R-A750EA 874 EUR
BOSSBS-0437R-A750
Anti-Streptavidin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.
Type: Primary
Antigen: Streptavidin
More Product Information