Anti-COX1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-0582R-CY3
BOSSBS-0582R-CY3EA 883 EUR
BOSSBS-0582R-CY3
Anti-COX1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
This is one of two genes encoding similar enzymes that catalyze the conversion of arachinodate to prostaglandin. The encoded protein regulates angiogenesis in endothelial cells, and is inhibited by nonsteroidal anti-inflammatory drugs such as aspirin. The protein may promote cell proliferation during tumor progression. Alternative splicing results in multiple transcript variants. [provided by RefSeq, Sep 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COX
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR