Anti-KIT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-0672R-HRP
BOSSBS-0672R-HRPEA 909 EUR
BOSSBS-0672R-HRP
Anti-KIT Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Tyrosine-protein kinase that acts as cell-surface receptor for the cytokine KITLG/SCF and plays an essential role in the regulation of cell survival and proliferation, hematopoiesis, stem cell maintenance, gametogenesis, mast cell development, migration and function, and in melanogenesis. In response to KITLG/SCF binding, KIT can activate several signaling pathways. Phosphorylates PIK3R1, PLCG1, SH2B2/APS and CBL. Activates the AKT1 signaling pathway by phosphorylation of PIK3R1, the regulatory subunit of phosphatidylinositol 3-kinase. Activated KIT also transmits signals via GRB2 andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol KIT
Environmentally Preferable
Antigen name Tyrosine-protein kinase Kit
Antigen synonyms Fdc|SOW3|CD117|SCO1|Gsfsow3|W|Ssm|Tr-kit|SCO5|Gsfsco5|Gsfsco1|Bs|c-KIT
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Dog,Rat,Mouse
Host Rabbit
Gene ID 3815
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Immunogen 928-964/988
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR