Anti-ANGPT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-0800R-A350
BOSSBS-0800R-A350EA 901 EUR
BOSSBS-0800R-A350
Anti-ANGPT1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Angiopoietins are proteins with important roles in vascular development and angiogenesis. All angiopoietins bind with similar affinity to an endothelial cell-specific tyrosine-protein kinase receptor. The protein encoded by this gene is a secreted glycoprotein that activates the receptor by inducing its tyrosine phosphorylation. It plays a critical role in mediating reciprocal interactions between the endothelium and surrounding matrix and mesenchyme and inhibits endothelial permeability. The protein also contributes to blood vessel maturation and stability, and may be involved in earlyMore Product Information