Anti-AVPR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-10014R-A488
BOSSBS-10014R-A488EA 893 EUR
BOSSBS-10014R-A488
Anti-AVPR2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Receptor for arginine vasopressin. The activity of this receptor is mediated by G proteins which activate adenylate cyclase. Involved in renal water reabsorption.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AVPR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR