Anti-SLC7A9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-10085R-HRP
BOSSBS-10085R-HRPEA 1350 EUR
BOSSBS-10085R-HRP
Anti-SLC7A9 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Involved in the high-affinity, sodium-independent transport of cystine and neutral and dibasic amino acids (system b(0,+)-like activity). Thought to be responsible for the high-affinity reabsorption of cystine in the kidney tubule.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: SLC7A9
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR