Anti-SLC7A5 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-10125R-CY5
BOSSBS-10125R-CY5EA 909 EUR
BOSSBS-10125R-CY5
Anti-SLC7A5 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Sodium-independent, high-affinity transport of large neutral amino acids such as phenylalanine, tyrosine, leucine, arginine and tryptophan, when associated with SLC3A2/4F2hc. Involved in cellular amino acid uptake. Acts as an amino acid exchanger. Involved in the transport of L-DOPA across the blood-brain barrier, and that of thyroid hormones triiodothyronine (T3) and thyroxine (T4) across the cell membrane in tissues such as placenta. Plays a role in neuronal cell proliferation (neurogenesis) in brain. Involved in the uptake of methylmercury (MeHg) when administered as the L-cysteine or D,L-More Product Information