Anti-MHC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-10251R-A647
BOSSBS-10251R-A647EA 893 EUR
BOSSBS-10251R-A647
Anti-MHC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol MHC1
Antigen name MHC Class I
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Flow cytometry Yes
Immunogen 60-105/181
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR