Anti-ENO2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-1027R-CY3
BOSSBS-1027R-CY3EA 901 EUR
BOSSBS-1027R-CY3
Anti-ENO2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Has neurotrophic and neuroprotective properties on a broad spectrum of central nervous system (CNS) neurons. Binds, in a calcium-dependent manner, to cultured neocortical neurons and promotes cell survival (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENO2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR