Anti-H3N1 hemagglutinin Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-10320R-CY5
BOSSBS-10320R-CY5EA 900 EUR
BOSSBS-10320R-CY5
Anti-H3N1 hemagglutinin Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Hemagglutinin (HA) is a class I viral fusion protein from Influenza virus. It is a major glycoprotein, comprising over 80% of the envelope proteins present in the virus particle. HA binds to sialic acid-containing receptors on the cell surface, bringing about the attachment of the virus particle to the cell, and is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane. The extent of infection into host organism is determined by HA. In natural infection, inactive HA is matured intoMore Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name H3N1 hemagglutinin
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Virus
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR