Anti-H3N1 hemagglutinin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
BS-10320R-HRP
BOSSBS-10320R-HRPEA 900 EUR
BOSSBS-10320R-HRP
Anti-H3N1 hemagglutinin Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Hemagglutinin (HA) is a class I viral fusion protein from Influenza virus. It is a major glycoprotein, comprising over 80% of the envelope proteins present in the virus particle. HA binds to sialic acid-containing receptors on the cell surface, bringing about the attachment of the virus particle to the cell, and is responsible for penetration of the virus into the cell cytoplasm by mediating the fusion of the membrane of the endocytosed virus particle with the endosomal membrane. The extent of infection into host organism is determined by HA. In natural infection, inactive HA is matured intoMore Product Information