Anti-PAF Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-1119R-A647
BOSSBS-1119R-A647EA 893 EUR
BOSSBS-1119R-A647
Anti-PAF Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Azelaoyl PAF is a potent PPARγ agonist which competes for the thiazolidinedione binding site.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAF
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR