Anti-JUNB Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-11228R-A350
BOSSBS-11228R-A350EA 921 EUR
BOSSBS-11228R-A350
Anti-JUNB Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Transcription factor involved in regulating gene activity following the primary growth factor response. Binds to the DNA sequence 5'-TGA[CG]TCA-3'.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JUNB
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR