Anti-NOVA1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-11324R-CY7
BOSSBS-11324R-CY7EA 900 EUR
BOSSBS-11324R-CY7
Anti-NOVA1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Nova-1 and Nova-2 are members of a superfamily of protein regulators of RNA metabolism in neurons. Both are nuclear RNA binding proteins with K homology motifs, conserved protein sequences which bind to RNA (1,2). Nova proteins, normally sequestered in the central nervous system, are expressed by systemic tumors in patients with the autoimmune disorder paraneoplastic opsoclonus-myoclonus ataxia (POMA) (3,4). Nova-1 is expressed in the hindbrain and ventral spinal cord and Nova-2 is expressed in the neocortex and hippocampus (4). Nova-1 is necessary for regulating neuron-specific alternativeMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol NOVA1
Antigen name Neuro-Oncological Ventral Antigen 1
Environmentally Preferable
Antigen synonyms leIF-F|IFN-alphaI|LeIF F|Nova-1
Antibody type Primary
Clonality Polyclonal
Conjugation Cy7®
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR