Anti-C1QC/C1QG Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
BS-11337R-CY7
BOSSBS-11337R-CY7EA 900 EUR
BOSSBS-11337R-CY7
Anti-C1QC/C1QG Rabbit Polyclonal Antibody (Cy7®)
Antibodies
C1q, a subcomponent of the classical complement pathway, is composed of nine subunits that mediate classical complement activation and thereby play an important role in the immune response. Six of these subunits are disulfide-linked dimers of chains A and B, while three of these subunits, designated C1q-A through C1q-C, are disulfide-linked dimers of chain C. The presence of receptors for C1q on effector cells modulates its activity, which may be antibody-dependent or independent. Macrophages are the primary source of C1q, while anti-inflammatory drugs as well as cytokines differentiallyMore Product Information