Anti-RIMS3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
BS-11359R-A750
BOSSBS-11359R-A750EA 890 EUR
BOSSBS-11359R-A750
Anti-RIMS3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Rab 3, a neural/neuroendocrine-specific member of the Rab family, is involved in Ca²⁺ regulated exocytosis and functions in an inhibitory capacity, controlling the recruitment and pooling of secretory vesicles at the plasma membrane. The Rim (Rab 3 interacting molecule) family of proteins (Rim1, Rim2, Rim3 and Rim4) are multidomain adaptors that regulate Rab 3 activity and sub-sequent neurotransmitter release. Rim3, also known as RIMS3 (regulating synaptic membrane exocytosis 3) or NIM3, is a 308 amino acid member of the Rim family. localised to the synapse and to cell junctions, Rim3More Product Information