Anti-ARG2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-11397R-A488
BOSSBS-11397R-A488EA 876 EUR
BOSSBS-11397R-A488
Anti-ARG2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
May play a role in the regulation of extra-urea cycle arginine metabolism and also in down-regulation of nitric oxide synthesis. Extrahepatic arginase functions to regulate L-arginine bioavailability to NO synthase. Since NO synthase is found in the penile corpus cavernosum smooth muscle, the clitoral corpus cavernosum and the vagina, arginase II plays a role in both male and female sexual arousal. It is therefore a potential target for the treatment of male and female sexual arousal disorders.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARG2
Clonality: Polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR