Anti-TFPI2 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-1144R
BOSSBS-1144REA 752 EUR
BOSSBS-1144R
Anti-TFPI2 Rabbit Polyclonal Antibody
Antibodies
May play a role in the regulation of plasmin-mediated matrix remodeling. Inhibits trypsin, plasmin, factor VIIa/tissue factor and weakly factor Xa. Has no effect on thrombin.

Recommended Dilutions: Western Blot: 1:100 to 1000; Flow Cytometry: 1:20 to 100; IHC-P: 1:100 to 500; IF(IHC-P): 1:50 to 200.

Type: Primary
Antigen: TFPI2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR