Anti-DCAMKL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-11455R-A555
BOSSBS-11455R-A555EA 893 EUR
BOSSBS-11455R-A555
Anti-DCAMKL1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Lissencephaly (smooth brain) is an abnormality of brain development characterized by incomplete neuronal migration and a smooth cerebral surface, manifesting as severe mental retardation. Genetic analysis has identified two proteins that are mutated in some cases of lissencephaly, designated lissencephaly-1 protein (LIS1) and doublecortin. LIS1 displays sequence homology to ?subunits of heterotrimeric G proteins, and doublecortin contains a consensus Abl phosphorylation site. In addition, the DCAMKL1 (doublecortin-like and CAM kinase-like 1) protein shows homology to doublecortin. All threeMore Product Information