Anti-HS1BP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-11568R-A350
BOSSBS-11568R-A350EA 893 EUR
BOSSBS-11568R-A350
Anti-HS1BP3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
HS1BP3 is a 392 amino acid protein that contains one PX (phox homology) domain, a leucine zipper, immunoreceptor tyrosine-based inhibitory motif-like motifs and multiple proline-rich regions. Expressed primarily in brain, HS1BP3 is encoded by a gene mapping to human chromosome 2p24.1. The gene encoding HS1BP3 is frequently mutated in familial essential tremor, a disorder characterized by kinetic tremor the the hands, voice or head, though there is no correlation to Parkinson disease. HS1BP3 interacts with HAX-1’s SH3 domain, and may also play a role in the regulation of catecholamine andMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HS1BP3
Environmentally Preferable
Antigen name HCLS1 binding protein 3
Antigen synonyms HS1-BP3|ETM2
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR