Anti-HAP40/F8a Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-11696R-A647
BOSSBS-11696R-A647EA 893 EUR
BOSSBS-11696R-A647
Anti-HAP40/F8a Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
The Huntingtin protein contains a polyglutamine region, which leads to Huntingtin’s disease (HD) when the number of glutamine repeats exceeds thirty-five. The mutated Huntingtin protein acts within the nucleus to induce neurodegeneration by a cell-specific apoptotic mechanism. The loss of activity of the Huntingtin protein may be contributed to abnormal interactions between the mutant protein and other associated cellular proteins. Huntingtin interacts with a variety of proteins including HAP1, glyceraldehyde phosphate dehydrogenase (GAPDH), HAP40, Rab5 and HIP1. HAP40 mediates theMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol HAP40/F8a
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR