Anti-CKAP1/TBCB Rabbit Polyclonal Antibody
Supplier: Bioss
BS-11713R
BOSSBS-11713REA 725 EUR
BOSSBS-11713R
Anti-CKAP1/TBCB Rabbit Polyclonal Antibody
Antibodies
Microtubules, the primary component of the cytoskeletal network, are highly dynamic structures composed of Alpha/Beta Tubulin heterodimers. Biosynthesis of functional microtubules involve the participation of several chaperones, termed Tubulin folding cofactors A (TBCA), B (TBCB), D (TBCD), E (TBCE) and C (TBCC), that act on folding intermediates downstream of the cytosolic chaperon, alternatively named TCP. TBCB (tubulin folding cofactor B), also known as CG22, CKAP1 or CKAPI, is a 244 amino acid cytoplasmic protein containing one CAP-Gly domain and in widely expressed. TBCB is involved inMore Product Information