Anti-ASH1L Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-11866R-HRP
BOSSBS-11866R-HRPEA 893 EUR
BOSSBS-11866R-HRP
Anti-ASH1L Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Histone methyltransferase specifically methylating 'Lys-36' of histone H3 (H3K36me).

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: ASH1L
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR