Anti-ZDHHC3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-12034R-A680
BOSSBS-12034R-A680EA 890 EUR
BOSSBS-12034R-A680
Anti-ZDHHC3 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Palmitoyltransferase with broad specificity. Palmitoylates GABA receptors on their gamma subunit (GABRG1, GABRG2 and GABRG3), which regulates synaptic clustering and/or cell surface stability. Palmitoylates glutamate receptors GRIA1 and GRIA2, which leads to their retention in Golgi. May also palmitoylate DLG4, DNAJC5 and SNAP25.

Type: Primary
Antigen: ZDHHC3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 201-299/299
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR